Methyl-2,5-dichloroisonicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2,5-dichloroisonicotinate, Methyl 2,5-dichloropyridine-4-carboxylate
048062-500MG
048062-1G
101882-634EA
184.62
USD
101882-634
101882-636
Methyl-2,5-dichloroisonicotinate ≥95%
Methyl 2,5-dichloroisonicotinate
Hazard: Irritant
Formula:
C₇H₅Cl₂NO₂ MW: 206.0300 g/mol Melting Pt: 28-29°C |
MDL Number:
MFCD09951944 CAS Number: 623585-74-0 |
Specification Test Results
Melting point | 28-29°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...