5-(4-Boc-piperazino)-2-nitroanisole ≥95%
Supplier: Matrix Scientific
048061-500MG
048061-1G
048061-5G
101882-628EA
150.77
USD
101882-628
101882-630
101882-632
5-(4-Boc-piperazino)-2-nitroanisole ≥95%
5-(4-Boc-piperazino)-2-nitroanisole
Hazard: Irritant
Formula:
C₁₆H₂₃N₃O₅ Melting Pt: 207…209 °C |
MDL Number:
MFCD09972251 CAS Number: 1017782-79-4 |
Specification Test Results
Melting point | 207-209°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...