1-(3-Methoxy-4-nitrophenyl)tetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

048060-500MG 048060-1G 048060-5G
101882-622EA 278.46 USD
101882-622 101882-624 101882-626
1-(3-Methoxy-4-nitrophenyl)tetrahydro-4(1H)-pyridinone ≥95%
1-(3-Methoxy-4-nitrophenyl)tetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄
Melting Pt: 134…135 °C
MDL Number: MFCD09972250
CAS Number: 761440-64-6

Order Now

Specification Test Results

Melting point 134-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR