1-(6-Chloro-2-methyl-4-pyrimidinyl)-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

048058-500MG 048058-1G 048058-5G
101882-610EA 375.38 USD
101882-610 101882-612 101882-614
1-(6-Chloro-2-methyl-4-pyrimidinyl)-4-piperidinecarboxylic acid ≥95%
1-(6-Chloro-2-methyl-4-pyrimidinyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₄ClN₃O₂
Melting Pt: 210…212 °C
MDL Number: MFCD09972249
CAS Number: 1017782-77-2

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR