2-{4-[(4-Fluorophenyl)sulfonyl]piperazino}-1-ethanol ≥95%

Supplier: Matrix Scientific

048057-500MG 048057-1G 048057-5G
101882-604EA 181.54 USD
101882-604 101882-606 101882-608
2-{4-[(4-Fluorophenyl)sulfonyl]piperazino}-1-ethanol ≥95%
2-{4-[(4-Fluorophenyl)sulfonyl]piperazino}-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₇FN₂O₃S
Melting Pt: 94…96 °C
MDL Number: MFCD01211992
CAS Number: 331845-78-4

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR