2-{4-[(4-Methylphenyl)sulfonyl]piperazino}-1-ethanol ≥95%

Supplier: Matrix Scientific

048056-500MG 048056-1G 048056-5G
101882-598EA 181.54 USD
101882-598 101882-600 101882-602
2-{4-[(4-Methylphenyl)sulfonyl]piperazino}-1-ethanol ≥95%
2-{4-[(4-Methylphenyl)sulfonyl]piperazino}-1-ethanol

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₃S
Melting Pt: 130…132 °C
MDL Number: MFCD00193707
CAS Number: 16017-64-4

Order Now

Specification Test Results

Melting point 130-132°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR