2-[4-(2-Amino-6-chloro-4-pyrimidinyl)piperazino]-1-ethanol ≥95%

Supplier: Matrix Scientific

048055-500MG 048055-1G 048055-5G
101882-592EA 278.46 USD
101882-592 101882-594 101882-596
2-[4-(2-Amino-6-chloro-4-pyrimidinyl)piperazino]-1-ethanol ≥95%
2-[4-(2-Amino-6-chloro-4-pyrimidinyl)piperazino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₆ClN₅O
Melting Pt: 179…°C180 °C
MDL Number: MFCD09972248
CAS Number: 1017782-76-1

Order Now

Specification Test Results

Melting point 179-°C180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR