2-{4-[(4-Iodophenyl)sulfonyl]piperazino}-1-ethanol ≥95%

Supplier: Matrix Scientific

048053-500MG 048053-1G 048053-5G
101882-582EA 181.54 USD
101882-582 101882-584 101882-586
2-{4-[(4-Iodophenyl)sulfonyl]piperazino}-1-ethanol ≥95%
2-{4-[(4-Iodophenyl)sulfonyl]piperazino}-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₇IN₂O₃S
Melting Pt: 120…122 °C
MDL Number: MFCD09761464
CAS Number: 941256-92-4

Order Now

Specification Test Results

Melting point 120-122°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR