2-{4-[(4-Chlorophenyl)sulfonyl]piperazino}-1-ethanol ≥95%

Supplier: Matrix Scientific

048052-500MG 048052-1G 048052-5G
101882-576EA 181.54 USD
101882-576 101882-578 101882-580
2-{4-[(4-Chlorophenyl)sulfonyl]piperazino}-1-ethanol ≥95%
2-{4-[(4-Chlorophenyl)sulfonyl]piperazino}-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃S
Melting Pt: 96…98 °C
MDL Number: MFCD01185744
CAS Number: 16017-65-5

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR