6-Chloro-2-methyl-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-4-hydroxy-2-methylpyrimidine, 6-Chloro-2-methylpyrimidin-4(1H)-one

048051-1G 048051-5G
101882-572EA 106.15 USD
101882-572 101882-574
6-Chloro-2-methyl-4-pyrimidinol ≥95%
6-Chloro-2-methyl-4-pyrimidinol

Hazard: Irritant

Formula: C₅H₅ClN₂O
MW: 144.558 g/mol
Melting Pt: 233…236 °C
MDL Number: MFCD00030773
CAS Number: 17551-52-9

Order Now

Specification Test Results

Melting point 233-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR