6-Chloro-N,2-dimethyl-4-pyrimidinamine ≥95%

Supplier: Matrix Scientific

048050-500MG 048050-1G 048050-5G
101882-566EA 153.85 USD
101882-566 101882-568 101882-570
6-Chloro-N,2-dimethyl-4-pyrimidinamine ≥95%
6-Chloro-N,2-dimethyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₆H₈ClN₃
MW: 157.6 g/mol
Melting Pt: 146…148 °C
MDL Number: MFCD09972247
CAS Number: 5621-01-2

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR