1-(3-Methoxy-4-nitrophenyl)-4-piperidinol ≥95%

Supplier: Matrix Scientific

048047-500MG 048047-1G 048047-5G
101882-548EA 147.69 USD
101882-548 101882-550 101882-552
1-(3-Methoxy-4-nitrophenyl)-4-piperidinol ≥95%
1-(3-Methoxy-4-nitrophenyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
Melting Pt: 139…140 °C
MDL Number: MFCD09972244
CAS Number: 761440-22-6

Order Now

Specification Test Results

Melting point 139-140°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR