1-(6-Chloro-2-methyl-4-pyrimidinyl)-4-piperidinol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(6-Chloro-2-methylpyrimidin-4-yl)-4-hydroxypiperidine

048046-500MG 048046-1G 048046-5G
101882-542EA 181.54 USD
101882-542 101882-544 101882-546
1-(6-Chloro-2-methyl-4-pyrimidinyl)-4-piperidinol ≥95%
1-(6-Chloro-2-methyl-4-pyrimidinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃O
MW: 227.7 g/mol
Melting Pt: 152…154 °C
MDL Number: MFCD09865027
CAS Number: 936845-82-8

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR