4-(4-Benzylpiperazino)-6-chloro-2-methylpyrimidine ≥95%
Supplier: Matrix Scientific
048045-500MG
048045-1G
048045-5G
101882-536EA
181.54
USD
101882-536
101882-538
101882-540
4-(4-Benzylpiperazino)-6-chloro-2-methylpyrimidine ≥95%
4-(4-Benzylpiperazino)-6-chloro-2-methylpyrimidine
Hazard: Irritant
Formula:
C₁₆H₁₉ClN₄ Melting Pt: 94…96 °C |
MDL Number:
MFCD09972243 CAS Number: 1017782-74-9 |
Specification Test Results
Melting point | 94-96°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...