4-Chloro-2-methyl-6-piperidinopyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-methyl-6-(piperidin-1-yl)pyrimidine

048044-500MG 048044-1G 048044-5G
101882-530EA 181.54 USD
101882-530 101882-532 101882-534
4-Chloro-2-methyl-6-piperidinopyrimidine ≥95%
4-Chloro-2-methyl-6-piperidinopyrimidine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MW: 211.7 g/mol
Melting Pt: 66…68 °C
MDL Number: MFCD00466217
CAS Number: 94052-15-0

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR