4-(6-Chloro-2-methylpyrimidin-4-yl)morpholine ≥95%

Supplier: Matrix Scientific

048042-500MG 048042-1G 048042-5G
101882-518EA 136.92 USD
101882-518 101882-520 101882-522
4-(6-Chloro-2-methylpyrimidin-4-yl)morpholine ≥95%
4-(6-Chloro-2-methylpyrimidin-4-yl)morpholine

Hazard: Irritant

Formula: C₉H₁₂ClN₃O
MW: 213.6600 g/mol
Melting Pt: 101…103 °C
MDL Number: MFCD07782031
CAS Number: 22177-99-7

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR