2-(4-Nitrophenyl)propanenitrile ≥95%
Supplier: Matrix Scientific
048040-1G
048040-5G
101882-508EA
110.77
USD
101882-508
101882-510
2-(4-Nitrophenyl)propanenitrile ≥95%
2-(4-Nitrophenyl)propanenitrile
Hazard: Irritant
Formula:
C₉H₈N₂O₂ MW: 176.18 g/mol Melting Pt: 78…80 °C |
MDL Number:
MFCD03427171 CAS Number: 50712-63-5 |
Specification Test Results
Melting point | 78-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...