Benzyl-N-[(3-chlorophenyl)(phenylsulfonyl)methyl]carbamate ≥95%
Supplier: Matrix Scientific
048037-500MG
048037-1G
048037-5G
101882-496EA
181.54
USD
101882-496
101882-498
101882-500
Benzyl-N-[(3-chlorophenyl)(phenylsulfonyl)methyl]carbamate ≥95%
Benzyl-N-[(3-chlorophenyl)(phenylsulfonyl)methyl]carbamate
Hazard: Irritant
Formula:
C₂₁H₁₈ClNO₄S MW: 415.9 g/mol Melting Pt: 159…161 °C |
MDL Number:
MFCD09972242 CAS Number: 1017782-73-8 |
Specification Test Results
Melting point | 159-161°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...