3-Methyl-1H-indazol-4-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-3-methyl-1H-indazole

048036-500MG 048036-1G
101882-492EA 707.69 USD
101882-492 101882-494
3-Methyl-1H-indazol-4-ol ≥95%
3-Methyl-1H-indazol-4-ol

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 197…199 °C
MDL Number: MFCD09027195
CAS Number: 149071-05-6

Order Now

Specification Test Results

Melting point 197-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR