4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

048034-500MG 048034-1G 048034-5G
101882-480EA 921.54 USD
101882-480 101882-482 101882-484
4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbaldehyde ≥95%
4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₇BrN₂O₂
Melting Pt: 129…131 °C
MDL Number: MFCD09152750
CAS Number: 952182-73-9

Order Now

Specification Test Results

Melting point 129-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR