N-(2-Bromo-4-fluorophenyl)thiourea ≥95%

Supplier: Matrix Scientific

048033-500MG 048033-1G
101882-476EA 246.15 USD
101882-476 101882-478
N-(2-Bromo-4-fluorophenyl)thiourea ≥95%
N-(2-Bromo-4-fluorophenyl)thiourea

Hazard: Irritant

Formula: C₇H₆BrFN₂S
MW: 249.11 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD09027194
CAS Number: 952182-71-7

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR