cis-3,5-Dimethyldihydro-2H-pyran-2,6(3H)-dione ≥95%

Supplier: Matrix Scientific

048031-500MG 048031-1G
101882-466EA 356.92 USD
101882-466 101882-468
cis-3,5-Dimethyldihydro-2H-pyran-2,6(3H)-dione ≥95%
cis-3,5-Dimethyldihydro-2H-pyran-2,6(3H)-dione

Hazard: Irritant

Formula: C₇H₁₀O₃
Melting Pt: 92…94 °C
MDL Number: MFCD09152749
CAS Number: 4295-92-5

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR