3-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

048030-500MG 048030-1G 048030-5G
101882-460EA 921.54 USD
101882-460 101882-462 101882-464
3-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbaldehyde ≥95%
3-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₇BrN₂O₂
Melting Pt: 86…88 °C
MDL Number: MFCD09152748
CAS Number: 952182-70-6

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR