4-((5-Bromopyrimidin-2-yl)oxy)aniline ≥95%

Supplier: Matrix Scientific

048028-500MG 048028-1G
101882-456EA 921.54 USD
101882-456 101882-458
4-((5-Bromopyrimidin-2-yl)oxy)aniline ≥95%
4-((5-Bromopyrimidin-2-yl)oxy)aniline

Hazard: Irritant

Formula: C₁₀H₈BrN₃O
MW: 266.0900 g/mol
Melting Pt: 67…69 °C
MDL Number: MFCD09152747
CAS Number: 76660-37-2

Order Now

Specification Test Results

Melting point 67-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR