5-Aminoimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: Imidazo[1,2-a]pyridin-5-amine

048025-500MG 048025-1G
101882-446EA 75.38 USD
101882-446 101882-448
5-Aminoimidazo[1,2-a]pyridine ≥95%
5-Aminoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₇N₃
MW: 133.1506 g/mol
Melting Pt: 156-158°C
MDL Number: MFCD05863377
CAS Number: 66358-23-4

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR