5-(Ethoxycarbonyl)-6-hydroxy-3-pyridinesulfonicacid ≥95%
Supplier: Matrix Scientific
048023-500MG
048023-1G
048023-5G
101882-440EA
278.46
USD
101882-440
101882-442
101882-444
5-(Ethoxycarbonyl)-6-hydroxy-3-pyridinesulfonicacid ≥95%
5-(Ethoxycarbonyl)-6-hydroxy-3-pyridinesulfonicacid
Hazard: Irritant
Formula:
C₈H₉NO₆S MW: 247.23 g/mol Melting Pt: 238…240 °C |
MDL Number:
MFCD09027192 CAS Number: 334708-06-4 |
Specification Test Results
Melting point | 238 - 240 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...