Ethyl-4-(4-methoxybenzoyl)-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

048019-500MG 048019-1G 048019-5G
101882-422EA 115.38 USD
101882-422 101882-424 101882-426
Ethyl-4-(4-methoxybenzoyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(4-methoxybenzoyl)-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₃NO₅
Melting Pt: 55…57 °C
MDL Number: MFCD09027191
CAS Number: 952182-69-3

Order Now

Specification Test Results

Melting point 55-57°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR