4-Chloro-N-[2-(hydroxymethyl)phenyl]benzenesulfonamide ≥95%
Supplier: Matrix Scientific
048018-500MG
048018-1G
048018-5G
101882-416EA
181.54
USD
101882-416
101882-418
101882-420
4-Chloro-N-[2-(hydroxymethyl)phenyl]benzenesulfonamide ≥95%
4-Chloro-N-[2-(hydroxymethyl)phenyl]benzenesulfonamide
Hazard: Irritant
Formula:
C₁₃H₁₂ClNO₃S Melting Pt: 145…147 °C |
MDL Number:
MFCD01647380 CAS Number: 353254-99-6 |
Specification Test Results
Melting point | 145-147°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...