5-[(4-Bromophenyl)sulfanyl]-1,3-thiazol-2-ylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-5-[(4-bromophenyl)thio]-1,3-thiazole
048017-500MG
048017-1G
048017-5G
101882-410EA
375.38
USD
101882-410
101882-412
101882-414
5-[(4-Bromophenyl)sulfanyl]-1,3-thiazol-2-ylamine ≥95%
5-[(4-Bromophenyl)sulfanyl]-1,3-thiazol-2-ylamine
Hazard: Irritant
Formula:
C₉H₇BrN₂S₂ MW: 287.21 g/mol Melting Pt: 174…176 °C |
MDL Number:
MFCD09027190 CAS Number: 90484-42-7 |
Specification Test Results
Melting point | 174-176°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...