2-Piperidino-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Piperidin-1-yl)thiazole-4-carboxylic acid

048016-500MG 048016-1G
101882-406EA 375.38 USD
101882-406 101882-408
2-Piperidino-1,3-thiazole-4-carboxylic acid ≥95%
2-Piperidino-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₂S
MW: 212.27 g/mol
Melting Pt: 186…188 °C
MDL Number: MFCD09027189
CAS Number: 952182-68-2

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR