5-Amino-6-bromo-2-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-6-bromopicolinonitrile

048015-500MG 048015-1G
101882-402EA 356.92 USD
101882-402 101882-404
5-Amino-6-bromo-2-pyridinecarbonitrile ≥95%
5-Amino-6-bromo-2-pyridinecarbonitrile

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.03 g/mol
Melting Pt: 172…174 °C
MDL Number: MFCD09027188
CAS Number: 849353-22-6

Order Now

Specification Test Results

Melting point 172-174°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR