2-((4-Chlorophenyl)amino)acetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Chloroanilino)-1-phenylethan-1-one
048013-500MG
048013-1G
048013-5G
101882-392EA
181.54
USD
101882-392
101882-394
101882-396
2-((4-Chlorophenyl)amino)acetophenone ≥95%
2-((4-Chlorophenyl)amino)acetophenone
Hazard: Irritant
Formula:
C₁₄H₁₂ClNO Melting Pt: 164-166°C |
MDL Number:
MFCD02028817 CAS Number: 53181-22-9 |
Specification Test Results
Melting point | 164-166°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...