1-Phenyl-2-[3-(trifluoromethyl)anilino]-1-ethanone ≥95%

Supplier: Matrix Scientific

048012-500MG 048012-1G 048012-5G
101882-386EA 181.54 USD
101882-386 101882-388 101882-390
1-Phenyl-2-[3-(trifluoromethyl)anilino]-1-ethanone ≥95%
1-Phenyl-2-[3-(trifluoromethyl)anilino]-1-ethanone

Hazard: Irritant

Formula: C₁₅H₁₂F₃NO
Melting Pt: 138…140 °C
MDL Number: MFCD09152744
CAS Number: 952182-66-0

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR