2-Amino-4-(4-nitrophenyl)-3-furonitrile ≥95%

Supplier: Matrix Scientific

048011-500MG 048011-1G 048011-5G
101882-380EA 356.92 USD
101882-380 101882-382 101882-384
2-Amino-4-(4-nitrophenyl)-3-furonitrile ≥95%
2-Amino-4-(4-nitrophenyl)-3-furonitrile

Hazard: Irritant

Formula: C₁₁H₇N₃O₃
Melting Pt: 210…212 °C
MDL Number: MFCD09027186
CAS Number: 606099-85-8

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR