2-((4-Bromophenyl)amino)acetophenone ≥95%

Supplier: Matrix Scientific

048010-500MG 048010-1G 048010-5G
101882-374EA 181.54 USD
101882-374 101882-376 101882-378
2-((4-Bromophenyl)amino)acetophenone ≥95%
2-((4-Bromophenyl)amino)acetophenone

Hazard: Irritant

Formula: C₁₄H₁₂BrNO
Melting Pt: 158-160°C
MDL Number: MFCD00598456
CAS Number: 4831-21-4

Order Now

Specification Test Results

Melting point 158-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR