2-(4-Methoxyanilino)-1,3-thiazole-4-carboxylicacid hydrobromide ≥95%
Supplier: Matrix Scientific
048005-500MG
048005-1G
048005-5G
101882-354EA
375.38
USD
101882-354
101882-356
101882-358
2-(4-Methoxyanilino)-1,3-thiazole-4-carboxylicacid hydrobromide ≥95%
2-(4-Methoxyanilino)-1,3-thiazole-4-carboxylicacid hydrobromide
Hazard: Irritant
Formula:
C₁₁H₁₁BrN₂O₃S Melting Pt: 235…237 °C |
MDL Number:
MFCD09027184 CAS Number: 1134600-62-6 |
Specification Test Results
Melting point | 235-237°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...