2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(3-Fluorophenyl)thiazole-4-carboxylic acid

048002-500MG 048002-1G
101882-346EA 747.69 USD
101882-346 101882-348
2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid ≥95%
2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆FNO₂S
MW: 223.23 g/mol
Melting Pt: 211…213 °C
MDL Number: MFCD07375293
CAS Number: 886369-06-8

Order Now

Specification Test Results

Melting point 211-213°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR