3-Bromo-4-[(4-fluorobenzyl)oxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047995-500MG 047995-1G 047995-5G
101882-316EA 181.54 USD
101882-316 101882-318 101882-320
3-Bromo-4-[(4-fluorobenzyl)oxy]benzenecarbaldehyde ≥95%
3-Bromo-4-[(4-fluorobenzyl)oxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀BrFO₂
Melting Pt: 139…141 °C
MDL Number: MFCD02605292
CAS Number: 6481-37-4

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR