3-[4-(Benzyloxy)-3-bromophenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

047991-500MG 047991-1G 047991-5G
101882-302EA 278.46 USD
101882-302 101882-304 101882-306
3-[4-(Benzyloxy)-3-bromophenyl]acrylic acid ≥95%
3-[4-(Benzyloxy)-3-bromophenyl]acrylic acid

Hazard: Irritant

Formula: C₁₆H₁₃BrO₃
Melting Pt: 215…218 °C
MDL Number: MFCD11841021
CAS Number: 67808-76-8

Order Now

Specification Test Results

Melting point 215-218°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR