6-Chloro-2-methyl[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one ≥95%

Supplier: Matrix Scientific

047987-500MG 047987-1G
101882-288EA 356.92 USD
101882-288 101882-290
6-Chloro-2-methyl[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one ≥95%
6-Chloro-2-methyl[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one

Hazard: Irritant

Formula: C₆H₅ClN₄O
Melting Pt: 174…176 °C
MDL Number: MFCD11553072
CAS Number: 65866-54-8

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR