5-Bromo-6-cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

047986-500MG 047986-1G
101882-284EA 538.46 USD
101882-284 101882-286
5-Bromo-6-cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
5-Bromo-6-cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₉H₇BrN₂O
MW: 239.08 g/mol
Melting Pt: 239…231 °C
MDL Number: MFCD11841020
CAS Number: 1135283-57-6

Order Now

Specification Test Results

Melting point 239-231°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR