(5-Amino-2-morpholinophenyl)methanol ≥95%

Supplier: Matrix Scientific

047985-500MG 047985-1G
101882-280EA 353.85 USD
101882-280 101882-282
(5-Amino-2-morpholinophenyl)methanol ≥95%
(5-Amino-2-morpholinophenyl)methanol

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
Melting Pt: 133…135 °C
MDL Number: MFCD11841019
CAS Number: 759456-60-5

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR