Ethyl-6-phenylimidazo[2,1-b][1,3]thiazole-2-carboxylate hydrobromide ≥95%

Supplier: Matrix Scientific

047982-500MG 047982-1G 047982-5G
101882-264EA 278.46 USD
101882-264 101882-266 101882-268
Ethyl-6-phenylimidazo[2,1-b][1,3]thiazole-2-carboxylate hydrobromide ≥95%
Ethyl-6-phenylimidazo[2,1-b][1,3]thiazole-2-carboxylate hydrobromide

Hazard: Irritant

Formula: C₁₄H₁₃BrN₂O₂S
Melting Pt: 220…222 °C
MDL Number: MFCD11553071
CAS Number: 1177093-01-4

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR