6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Cyclopropyl-2-hydroxynicotinonitrile

047981-500MG 047981-1G
101882-260EA 375.38 USD
101882-260 101882-262
6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₉H₈N₂O
MW: 160.18 g/mol
Melting Pt: 239…241 °C
MDL Number: MFCD11553070
CAS Number: 847144-72-3

Order Now

Specification Test Results

Melting point 239-241°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR