4-(Benzyloxy)-3-bromobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Benzyloxy-3-bromobenzaldehyde

047980-500MG 047980-1G 047980-5G
101882-254EA 181.54 USD
101882-254 101882-256 101882-258
4-(Benzyloxy)-3-bromobenzaldehyde ≥95%
4-(Benzyloxy)-3-bromobenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁BrO₂
MW: 291.1400 g/mol
Melting Pt: 96-99°C
MDL Number: MFCD02629356
CAS Number: 69455-12-5

Order Now

Specification Test Results

Melting point 96-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR