Methyl-2-bromo-4-morpholinobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

047978-500MG 047978-1G
101882-250EA 264.62 USD
101882-250 101882-252
Methyl-2-bromo-4-morpholinobenzenecarboxylate ≥95%
Methyl-2-bromo-4-morpholinobenzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₄BrNO₃
Melting Pt: 76…78 °C
MDL Number: MFCD11553069
CAS Number: 1092352-96-9

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR