[5-Amino-2-(4-methylpiperazino)phenyl]methanol ≥95%
Supplier: Matrix Scientific
047976-500MG
047976-1G
101882-242EA
361.54
USD
101882-242
101882-244
[5-Amino-2-(4-methylpiperazino)phenyl]methanol ≥95%
[5-Amino-2-(4-methylpiperazino)phenyl]methanol
Hazard: Irritant
Formula:
C₁₂H₁₉N₃O Melting Pt: 162…164 °C |
MDL Number:
MFCD11841016 CAS Number: 802541-81-7 |
Specification Test Results
Melting point | 162-164°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...