Methyl-5-amino-2-(4-methylpiperazino)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

047972-500MG 047972-1G 047972-5G
101882-228EA 375.38 USD
101882-228 101882-230 101882-232
Methyl-5-amino-2-(4-methylpiperazino)benzenecarboxylate ≥95%
Methyl-5-amino-2-(4-methylpiperazino)benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₂
Melting Pt: 89…91 °C
MDL Number: MFCD11841015
CAS Number: 774126-94-2

Order Now

Specification Test Results

Melting point 89-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR