3-(4-Bromophenyl)-5-(chloromethyl)-1,2,4-oxadiazole ≥95%
Supplier: Matrix Scientific
047971-500MG
047971-1G
047971-5G
101882-222EA
375.38
USD
101882-222
101882-224
101882-226
3-(4-Bromophenyl)-5-(chloromethyl)-1,2,4-oxadiazole ≥95%
3-(4-Bromophenyl)-5-(chloromethyl)-1,2,4-oxadiazole
Hazard: Irritant
Formula:
C₉H₆BrClN₂O Melting Pt: 63…65 °C |
MDL Number:
MFCD08445385 CAS Number: 110704-42-2 |
Specification Test Results
Melting point | 63-65°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...