8-Bromo-6-methoxy-2-methyl-2H-1,4-benzoxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

047969-500MG 047969-1G
101882-214EA 715.38 USD
101882-214 101882-216
8-Bromo-6-methoxy-2-methyl-2H-1,4-benzoxazin-3(4H)-one ≥95%
8-Bromo-6-methoxy-2-methyl-2H-1,4-benzoxazin-3(4H)-one

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₃
Melting Pt: 183…185 °C
MDL Number: MFCD11553068
CAS Number: 1092352-94-7

Order Now

Specification Test Results

Melting point 183-185°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR